ContactUsLinkComponent
Distilling heads, Claisen type
Distilling heads, Claisen type
Catalog # WITG1.020.004
Supplier:  witeg Labortechnik
undefined
Distilling heads, Claisen type
Catalog # WITG1.020.004
Supplier:  witeg Labortechnik
Specifications
  • Cone to condenser:
    19/26 NS
  • Cone to flask:
    24/29 NS
  • Pk:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Acc. to DIN 12594 and in addition to standard, with two parallel NS sockets and two NS cones (descending angle: 75°), for distillation thermometers of 55 mm built-in length.