ContactUsLinkComponent
Reusable earplugs, Rockets®
Reusable earplugs, Rockets®
Catalog # 111-0720
Supplier:  Moldex
undefined
Reusable earplugs, Rockets®
Catalog # 111-0720
Supplier:  Moldex
Specifications
  • Colour:
    Blue
  • Description:
    Earplugs, with cord and box
  • Size:
    U*
  • Pk:
    50 PAIR
Specifications
About this item
Corded earplugs made from soft TPE material which is gentle on the skin. The flexible design and the solid handle ensures easy insertion and removal of plugs.
  • Fully detectable
  • Air-cushioned tip
  • Comes with handy PocketPak storage container
Washable and reusable, ideal for the food industry.
SNR: 27 dB
Certifications: EN 352-2:2002
Caution: * One size fits all.