Orders LinkContactUsLinkComponent
Measuring cylinders, conical
Measuring cylinders, conical
Measuring cylinders, conical
Certificates
Measuring cylinders, conical
Measuring cylinders, conical
2 Options Available Below
Product Details & Documents
PP, translucent.
  • Good chemical resistance
  • Autoclavable to +121 °C
  • Liquid volume is easy to read
Ideal for a wide range of applications. With moulded graduations. Broad base for stability.
Product Family Options
Product Information
  • Capacity
    Division
    Height
    Availability
    Price
  • 500 ml
    10 ml
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 12,10 €
    12,10 €
    1 * 1 ST
       12,10 €
  • 250 ml
    5 ml
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,80 €
    9,80 €
    1 * 1 ST
       9,80 €