Sudan black B
Catalog # EHERC16986110
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:0,1 g
- Seq:3020
Specifications
About this item
:
:456.55 g/mol
Storage Temperature:Ambient
EINECS:224-087-1
CAS Number:4197-25-5
Formula:C₂₉H₂₄N₆
MDL Number:MFCD00006919