Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,2-Dimethyl-1,1,2,2-tetraphenyldisilane
1,2-Dimethyl-1,1,2,2-tetraphenyldisilane
1,2-Dimethyl-1,1,2,2-tetraphenyldisilane
SILNSILS07600.5
:
CHEMPUR SILANE
:
1,2-Dimethyl-1,1,2,2-tetraphenyldisilane
SILNSILS07600.5
:
CHEMPUR SILANE
:
Pack type:
Glass bottle for solids
Pk:
5 g
Seq:
3173
:
:
394.6557 g/mol
:
214-632-1
:
1172-76-5
:
C₂₆H₂₆Si₂
:
MFCD00051536