Order Entry
Germany
ContactUsLinkComponent
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) substrate, powder for ELISA
Irritant
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) substrate, powder for ELISA
Catalog # SOUT0202-01
CAS Number:  
Irritant
ABTS (Diammonium 2,2'-azinobis[3-ethyl-2,3-dihydrobenzothiazole-6-sulphonate]) substrate, powder for ELISA
Catalog # SOUT0202-01
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :548.68 g/mol
EINECS:250-396-6
 :GHS07
Storage Temperature:Refrigerator
CAS Number:30931-67-0
Formula:C₁₈H₂₄N₆O₆S₄
MDL Number:MFCD00010404