EPPS (HEPPS, 4-(2-hydroxyethyl)-1-piperazinepropanesulphonic acid)
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:16052-06-5
:
Formula:C₉H₂₀N₂O₄S
MDL Number:MFCD00006160
:252.33 g/mol
EINECS:240-198-8
:GHS07
Storage Temperature:Ambient