Order Entry
Germany
ContactUsLinkComponent
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
Certificates
Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)

Specifications

  • CAS No.:
    58970-76-6
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00083262
  • Protein synonyms:
    Ubenimex|Bestatin-S-R-S-N-3-amino-2-hydroxy-4-phenylbutyroyl-L-leucine|N-[(2S,3R)-3-Amino-2-hydroxy-4-phenylbutyryl]-L-leucine
  • Protein/peptide name:
    Bestatin ([S-(R*,S*)]-N-(3-amino-2-hydroxy-4-phenylbutyroyl)-L-leucine)
  • Molecular weight:
    308,4 kDa
  • Molecular formula:
    C₁₆H₂₄N₂O₄

Specifications