Acriflavine hydrochloride fluorescent dye, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:8063-24-9
:69235-50-3
Formula:C₂₇H₂₈Cl₄N₆
MDL Number:MFCD00069039
Melting Pt:260 °C (dec., lit.)
ADR:9,III
:M-Clarity™ quality level = MQ200
:578.36 g/mol
UN:3077
:GHS05
:GHS07
:GHS09
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Faint Brown to Brown and Orange to Dark Orange and Red to Very Dark Red
- Appearance (Form):Powder
- Solubility (Colour):Orange to Brown
- Solubility (Turbidity)
1 mg/ml, H₂O:Clear - Water (by Karl Fischer):≤13 %
- Infrared spectrum:Conforms to Structure
- % Acriflavine HCL content:10 - 50
- % Proflavine HCL:25 - 70
- Recommended Retest Period:3 years