Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Undecyl-β-D-maltoside
n-Undecyl-β-D-maltoside
n-Undecyl-β-D-maltoside
n-Undecyl-β-D-maltoside
Supplier:
BIOMOL RESEARCH LABORATORIES
Certificates
Expand image
n-Undecyl-β-D-maltoside
Supplier:
BIOMOL RESEARCH LABORATORIES
About this item
CAS Number:
170552-39-3
:
Formula:
C₂₃H₄₄O₁₁