n-Nonyl-β-D-maltoside
Catalog # MOLE61253204-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:468.54 g/mol
CAS Number:106402-05-5
Formula:C₂₁H₄₀O₁₁
MDL Number:MFCD08062400