Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Nonyl-β-D-maltoside
n-Nonyl-β-D-maltoside
n-Nonyl-β-D-maltoside
n-Nonyl-β-D-maltoside
Supplier:
BIOMOL RESEARCH LABORATORIES
Certificates
Expand image
n-Nonyl-β-D-maltoside
Supplier:
BIOMOL RESEARCH LABORATORIES
About this item
CAS Number:
106402-05-5
:
Formula:
C₂₁H₄₀O₁₁
MDL Number:
MFCD08062400
:
468.54 g/mol