
n-Dodecyl-β-D-maltoside, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:69227-93-6
:
Formula:C₂₄H₄₆O₁₁
MDL Number:MFCD00043012
:510.6 g/mol
:M-Clarity™ quality level = MQ200
:GHS07
Storage Temperature:Freezer
Frequently Bought Together