Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside
Supplier:
Serva
Certificates
Expand image
n-Dodecyl-β-D-maltoside
Supplier:
Serva
About this item
CAS Number:
69227-93-6
:
Formula:
C₂₄H₄₆O₁₁
MDL Number:
MFCD00043012
:
510.6 g/mol
:
GHS07
Storage Temperature:
Freezer