
n-Dodecyl-β-D-maltoside
Catalog # MOLE21771928-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:510.6 g/mol
:GHS07
Storage Temperature:Freezer
CAS Number:69227-93-6
Formula:C₂₄H₄₆O₁₁
MDL Number:MFCD00043012