Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Dodecyl-β-D-maltoside
n-Dodecyl-β-D-maltoside, Millipore®
n-Dodecyl-β-D-maltoside, Millipore®
:
Merck Millipore (Calbiochem)
n-Dodecyl-β-D-maltoside, Millipore®
:
Merck Millipore (Calbiochem)
:
69227-93-6
:
:
C₂₄H₄₆O₁₁
:
MFCD00043012
:
510.6 g/mol
:
GHS07
:
Freezer