ContactUsLinkComponent
n-Dodecyl-β-D-maltoside 99%, high purity
Irritant
n-Dodecyl-β-D-maltoside 99%, high purity
Certificates
undefined
Irritant
n-Dodecyl-β-D-maltoside 99%, high purity
Specification Test Results
  • Appearance:
    White to off-white Powder
  • Infrared spectrum:
    Conforms
  • HPLC:
    ≥98.5 % (ELSD)
  • Specific optical rotation:
    +45° to +49.5° (20°C, 589 nm) (c=1, water)