Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
n-Decyl-β-D-maltoside
n-Decyl-β-D-maltoside, Sigma-Aldrich®
n-Decyl-β-D-maltoside, Sigma-Aldrich®
n-Decyl-β-D-maltoside, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
n-Decyl-β-D-maltoside, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
82494-09-5
:
Formula:
C₂₂H₄₂O₁₁
MDL Number:
MFCD00061624
:
482.56 g/mol
:
M-Clarity™ quality level = MQ100