Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Proteins and Peptides
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Soy bean trypsin inhibitor
Supplier:
Merck
Certificates
Expand image
Soy bean trypsin inhibitor
Supplier:
Merck
About this item
Specifications
CAS No.:
9035-81-8
Species:
Soy bean
Storage conditions:
Freezer
Protein/peptide name:
Trypsin inhibitor
Specifications