Orders LinkContactUsLinkComponent
Phosphoramidon
Phosphoramidon
10 mg
Catalog # 786-054
Supplier:   G-Biosciences
Phosphoramidon
Catalog # 786-054
Supplier:  G-Biosciences
CAS Number:  
Phosphoramidon
10 mg
CAS Number:  
Price
502,00 €
Log in for availability
1 * 10 mg
Product Details & Documents
Phosphoramidon inhibits some metalloproteases, including thermolysin, collagenase and bacterial metallo proteases from Bacillus subtilis, Streptomyces griseus and Pseudomonas aeruginosa (metallo elastase). Soluble in water, methanol and DMSO.
Specifications
Specifications
  • Pk:
    10 mg
  • Cat. No.:
    786-054
  • Protein/peptide name:
    Phosphoramidon
  • CAS No.:
    36357-77-4