0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Proteins and Peptides
E-64D
E-64D
UPTIUP827190
:
Uptima
:
E-64D
UPTIUP827190
:
Uptima
:
Pack type:
Glass bottle for solids
Pk:
5 mg
CAS No.:
88321-09-9
Protein synonyms:
(EST|3S)-(+)-trans-Epoxy succinyl-L-leucylamido-3-methyl butane ethyl ester)|(2S
Protein/peptide name:
E-64D