Adenosine-5'-triphosphate disodium salt (ATP disodium salt) hydrate, (max. 10% H₂O) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:34369-07-8
:
Formula:C₁₀H₁₄N₅Na₂O₁₃P₃·xH₂O
MDL Number:MFCD00150755
:551.14 g/mol (anhy)
Merck Index:14,155
Storage Temperature:Freezer
EINECS:213-579-1