Order Entry
Germany
ContactUsLinkComponent
Adenosine-5'-diphosphoric acid (ADP)
Adenosine-5'-diphosphoric acid (ADP)
Catalog # MSPPCH384
CAS Number:  
Adenosine-5'-diphosphoric acid (ADP)
Catalog # MSPPCH384
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :427.2 g/mol
EINECS:200-392-5
CAS Number:58-64-0
Formula:C₁₀H₁₅N₅O₁₀P₂
MDL Number:MFCD00066473