Adenosine-5'-diphosphoric acid (ADP) ≥96%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:58-64-0
:
Formula:C₁₀H₁₅N₅O₁₀P₂
MDL Number:MFCD00066473
:427.2 g/mol
Storage Temperature:Freezer
EINECS:200-392-5
Specification Test Results
- Identity Test:Passes
- Purity:≥96%
- FTIR:Conforms to Reference
- OD 250/OD 260:<1.0
- OD 280/OD 260:<0.5