8-Bromoadenosine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:2946-39-6
:
Formula:C₁₀H₁₂BrN₅O₄
MDL Number:MFCD00005733
:346.15 g/mol
Melting Pt:210…212 °C (dec.)
Storage Temperature:Refrigerator
EINECS:220-959-0
Specification Test Results
- Appearance:White to cream Crystalline powder
- Infrared spectrum:Conforms
- HPLC:≥97.5 %
- Specific optical rotation:-53° to -56° (20°C, 589 nm) (c=2.5, DMSO)
- NMR (1H-NMR):Consistent with structure