Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Guanosine 5'-triphosphate disodium salt
Guanosine 5'-triphosphate disodium salt
Guanosine 5'-triphosphate disodium salt
Guanosine 5'-triphosphate disodium salt
Supplier:
Serva
Certificates
Expand image
Guanosine 5'-triphosphate disodium salt
Supplier:
Serva
About this item
CAS Number:
56001-37-7
:
MFCD00083629
Formula:
C₁₀H₁₄N₅Na₂O₁₄P₃
MDL Number:
MFCD03410297
:
567.147 g/mol
EINECS:
259-940-7