Guanosine 5'-triphosphate disodium salt ≥95%, white powder
Supplier: MP Biomedicals
Certificates
About this item
CAS Number:56001-37-7
:MFCD00083629
Formula:C₁₀H₁₄N₅Na₂O₁₄P₃
MDL Number:MFCD03410297
:567.147 g/mol
Melting Pt:206…207 °C (lit.)
EINECS:259-940-7
:GHS07
Storage Temperature:Freezer
Specification Test Results
- Identity Test:Passes
- Purity:≥95%
- FTIR:Conforms to Standard
- UV/VIS: l max (water):252 ± 3nm
- OD250/OD260:1.20 ± 0.08
- OD280/OD260:0.65 ± 0.08
- OD290/OD260:0.23 ± 0.08