Order Entry
Germany
ContactUsLinkComponent
Chromotrope 2R suitable for modified Gomori trichrome stain, Sigma-Aldrich®
Chromotrope 2R suitable for modified Gomori trichrome stain, Sigma-Aldrich®
Certificates
undefined
Chromotrope 2R suitable for modified Gomori trichrome stain, Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Red to Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour), 1 mg/ml, H₂O:
    Dark Red
  • Nitrogen:
    4.3 - 7.6%
  • Sodium (Na), By ICP:
    ≥9%
  • Suitability, Suitable for use in modified Gomori Trichrome Stain:
    Suitable
  • E1%, in MeOH:
    ≥450
  • Wavelength:
    510 - 513 nm
  • Dye Content:
    ≥70%

Specification Test Results