CAPS sodium salt (3-(cyclohexylamino)propanesulphonic acid sodium salt)
Supplier: BIOMOL RESEARCH LABORATORIES
Certificates
About this item
CAS Number:105140-23-6
:
Formula:C₉H₁₈NNaO₃S
MDL Number:MFCD00269798
:243.3000 g/mol
Storage Temperature:Ambient