(2-Nitrophenyl)-β-D-galactopyranoside (ONPG)
Catalog # UPTIUP556683
Supplier: Uptima
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:301.25 g/mol
Storage Temperature:Freezer
EINECS:206-716-1
CAS Number:369-07-3
Formula:C₁₂H₁₅NO₈
MDL Number:MFCD00063255