Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Erythromycin
Erythromycin, Sigma-Aldrich®
Erythromycin, Sigma-Aldrich®
Erythromycin, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Erythromycin, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
114-07-8
:
Formula:
C₃₇H₆₇NO₁₃
MDL Number:
MFCD00084654
:
733.95 g/mol
:
M-Clarity™ quality level = MQ200
EINECS:
204-040-1
:
GHS07
Storage Temperature:
Ambient