Order Entry
Germany
ContactUsLinkComponent
Erythromycin
Erythromycin
Catalog # EHERC13203490
Supplier:  EHRENSTORFER
CAS Number:  
Erythromycin
Catalog # EHERC13203490
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,1 g
  • Seq:
    3020

Specifications

About this item

 :
 :733.95 g/mol
EINECS:204-040-1
 :GHS07
Storage Temperature:Ambient
CAS Number:114-07-8
Formula:C₃₇H₆₇NO₁₃
MDL Number:MFCD00084654