Order Entry
Germany
ContactUsLinkComponent
 
Carbenicillin disodium salt, free flowing powder
Catalog # ICNA195092.2
Supplier:  MP Biomedicals
CAS Number:  
Carbenicillin disodium salt, free flowing powder
Catalog # ICNA195092.2
Supplier:  MP Biomedicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 mg
  • Seq:
    2590

Specifications

Specification Test Results

  • Identity Test :
    Passes
  • Potency:
    >650 µg/mg

Specification Test Results

About this item

 :
 :422.363 g/mol
EINECS:225-360-8
 :GHS07
 :GHS08
Storage Temperature:Refrigerator
CAS Number:4800-94-6
Formula:C₁₇H₁₆N₂O₆S·2Na
MDL Number:MFCD00077683