(±)-Tetrahydrofurfuryl chloride
Catalog # MOLE34755704-100G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:120.58 g/mol
EINECS:221-102-3
CAS Number:3003-84-7
Formula:C₅H₉ClO
MDL Number:MFCD00005370