Order Entry
Germany
Orders LinkContactUsLinkComponent
Streptavidin

Streptavidin

Supplier:   Uptima
Streptavidin
Supplier:  Uptima
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • UPTIUP51558B
  • UPTIUP51558E
  • Protein/peptide name:
  • Streptavidin
  • Streptavidin
  • CAS No.:
  • 9013-20-1
  • 9013-20-1
  • MDL No.:
  • MFCD00082035
  • MFCD00082035
  • Storage conditions:
  • Freezer
  • Freezer

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    2 mg
    Log in for availability
    Log in for availability
    1 * 2 mg
    1 * 2 mg

    Specifications:
    More
    Specifications
    Cat. No.
    UPTIUP51558B
    Protein/peptide name
    Streptavidin
    CAS No.
    9013-20-1
    MDL No.
    MFCD00082035
    Storage conditions
    Freezer
  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg

    Specifications:
    More
    Specifications
    Cat. No.
    UPTIUP51558E
    Protein/peptide name
    Streptavidin
    CAS No.
    9013-20-1
    MDL No.
    MFCD00082035
    Storage conditions
    Freezer