Order Entry
Germany
ContactUsLinkComponent
Pluronic® L-121 (Synperonic® L121), Sigma-Aldrich®
Pluronic® L-121 (Synperonic® L121), Sigma-Aldrich®
Catalog # SIAL435457-250ML
Supplier:  Merck
CAS Number:  
Pluronic® L-121 (Synperonic® L121), Sigma-Aldrich®
Catalog # SIAL435457-250ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Seq:
    2590

Specifications

About this item

 :
 :4400 g/mol
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
CAS Number:9003-11-6