Order Entry
Germany
ContactUsLinkComponent
Pluronic® F-68, BioChemica
Pluronic® F-68, BioChemica
Catalog # A1288.0500
Supplier:  PanReac AppliChem
CAS Number:  
Pluronic® F-68, BioChemica
Catalog # A1288.0500
Supplier:  PanReac AppliChem
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :8400
Storage Temperature:Ambient
CAS Number:9003-11-6
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number:MFCD00082049