Pluronic® F-108 ~Mn 14,600, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:9003-11-6
:
Formula:HO(C₂H₄O)a(C₃H₆O)b(C₂H₄O)a
MDL Number:MFCD00082049
:14600 g/mol
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient