
TCEP-HCl (Tris(2-carboxyethyl)phosphine hydrochloride)
Catalog # UPTIUP242214
Supplier: Uptima
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:286.65 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
:GHS05
CAS Number:51805-45-9
Formula:C₉H₁₆ClO₆P
MDL Number:MFCD00145469