Order Entry
Germany
Orders LinkContactUsLinkComponent
Aprotinin

Aprotinin

Supplier:   Uptima
Aprotinin
Supplier:  Uptima
Certificates

2 Options Available Below

Product Details & Documents
Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • UPTIUP185582
  • UPTIUP185586
  • Protein/peptide name:
  • Aprotinin
  • Aprotinin
  • Protein synonyms:
  • Trypsin inhibitor, pancreatic basic
  • Trypsin inhibitor, pancreatic basic
  • CAS No.:
  • 9087-70-1
  • 9087-70-1
  • MDL No.:
  • MFCD00130541
  • MFCD00130541
  • Molecular formula:
  • C₂₈₄H₄₃₂N₈₄O₇₉S₇
  • C₂₈₄H₄₃₂N₈₄O₇₉S₇
  • Storage conditions:
  • Refrigerator
  • Refrigerator

Specifications

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 mg
    Log in for availability
    Log in for availability
    1 * 10 mg
    1 * 10 mg

    Specifications:
    More
    Specifications
    Cat. No.
    UPTIUP185582
    Protein/peptide name
    Aprotinin
    Protein synonyms
    Trypsin inhibitor, pancreatic basic
    CAS No.
    9087-70-1
    MDL No.
    MFCD00130541
    Molecular formula
    C₂₈₄H₄₃₂N₈₄O₇₉S₇
    Storage conditions
    Refrigerator
  • Glass bottle for solids
    50 mg
    Log in for availability
    Log in for availability
    1 * 50 mg
    1 * 50 mg

    Specifications:
    More
    Specifications
    Cat. No.
    UPTIUP185586
    Protein/peptide name
    Aprotinin
    Protein synonyms
    Trypsin inhibitor, pancreatic basic
    CAS No.
    9087-70-1
    MDL No.
    MFCD00130541
    Molecular formula
    C₂₈₄H₄₃₂N₈₄O₇₉S₇
    Storage conditions
    Refrigerator