Order Entry
Germany
ContactUsLinkComponent
N-Acetylglycine (Ac-Gly-OH)
N-Acetylglycine (Ac-Gly-OH)
Catalog # MOLEM76007738
Supplier:  Molekula
CAS Number:  
N-Acetylglycine (Ac-Gly-OH)
Catalog # MOLEM76007738
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g
  • Seq:
    3645

Specifications

About this item

 :
 :117.1 g/mol
EINECS:208-839-6
CAS Number:543-24-8
Formula:C₄H₇NO₃
MDL Number:MFCD00004275