3-Hydroxy-4-iodobenzaldehyde
Marketsource
Supplier: CHEMPUR
About this item
- CAS Number:135242-71-6
- Formula:C₇H₅IO₂
- MDL Number:MFCD00114697
- Molecular Weight:248.0179 g/mol
2 Options Available Below
Product Details & Documents
- CAS Number:135242-71-6
- Formula:C₇H₅IO₂
- MDL Number:MFCD00114697
- Molecular Weight:248.0179 g/mol
Specifications for Product Family
Specifications
Product Family Options
Product Information
- Pack typePkAvailabilityPrice
- Specifications:
More
Specifications
Cat. No.CHMPBTB10430.250 - Specifications:Marketsource
More
Specifications
Cat. No.MAYBBTB10430DAMarketsource
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



