Order Entry
Germany
ContactUsLinkComponent
Biebrich scarlet (contains ~60% dye content) , Sigma-Aldrich®
Biebrich scarlet (contains ~60% dye content) , Sigma-Aldrich®
Certificates
undefined
Biebrich scarlet (contains ~60% dye content) , Sigma-Aldrich®

Specification Test Results

  • Appearance (Colour):
    Red to Brown
  • Appearance (Form):
    Powder
  • Solubility (Colour):
    Orange to Red
  • Solubility (Turbidity), 1 mg/ml, H₂O:
    Clear to Slightly Hazy
  • Infrared spectrum:
    Conforms to Structure
  • Carbon:
    ≥22.5%
  • Nitrogen:
    ≥4.7%
  • Dye Content:
    ≥55%
  • EMM, Lambda max 510 nm:
    ≥19000

Specification Test Results