Order Entry
Germany
Orders LinkContactUsLinkComponent
 

Biebrich scarlet

Glass bottle for solids, 100 g
Catalog # MOLEM78385699
Marketsource
Supplier:   Molekula
Biebrich scarlet
Catalog # MOLEM78385699
Supplier:  Molekula
Marketsource
CAS Number:  4196-99-0
Biebrich scarlet

Glass bottle for solids, 100 g

CAS Number:  4196-99-0
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  Bonceau BS, C.I. 26905, Acid Red 66, Double Scarlet BSF, Croceine Scarlet, Biebrich Scarlet sodium salt
  • CAS Number:4196-99-0
  • Formula:C₂₂H₁₄N₄Na₂O₇S₂
  • MDL Number:MFCD00003891
  • Molecular Weight:556.49 g/mol
  • Storage Temperature:Ambient
  • EINECS:224-084-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g

Specifications