Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tiglic acid
Tiglic acid, Sigma-Aldrich®
Tiglic acid, Sigma-Aldrich®
Tiglic acid, Sigma-Aldrich®
SIALT35203-100G
:
Merck
:
Tiglic acid, Sigma-Aldrich®
SIALT35203-100G
:
Merck
:
Pack type:
Glass bottle for solids
Pk:
100 g
Seq:
3401
:
:
100.12 g/mol
:
8,III
:
M-Clarity™ quality level = MQ200
:
201-295-0
:
3261
:
80-59-1
:
C₅H₈O₂
:
MFCD00066864