Order Entry
Germany
ContactUsLinkComponent
1,3-Diisopropylimidazolium chloride ≥97%
1,3-Diisopropylimidazolium chloride ≥97%
Catalog # STRE07-0485-1G
CAS Number:  
1,3-Diisopropylimidazolium chloride ≥97%
Catalog # STRE07-0485-1G
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :188.70 g/mol
 :GHS07
CAS Number:139143-09-2
Formula:C₉H₁₇ClN₂
MDL Number:MFCD03840577