Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,3-Diisopropylimidazolium chloride
1,3-Diisopropylimidazolium chloride, Sigma-Aldrich®
1,3-Diisopropylimidazolium chloride, Sigma-Aldrich®
1,3-Diisopropylimidazolium chloride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
1,3-Diisopropylimidazolium chloride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
139143-09-2
:
Formula:
C₉H₁₇ClN₂
MDL Number:
MFCD03840577
:
M-Clarity™ quality level = MQ200
:
188.70 g/mol