Order Entry
Germany
ContactUsLinkComponent
Dodecyltrimethylammonium chloride 25% in aqueous solution
Irritant
Environment
Dodecyltrimethylammonium chloride 25% in aqueous solution
Catalog # MOLE54952018-1L
Supplier:  Molekula
CAS Number:  
Irritant
Environment
Dodecyltrimethylammonium chloride 25% in aqueous solution
Catalog # MOLE54952018-1L
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :263.89 g/mol
EINECS:203-927-0
 :GHS07
 :GHS09
Storage Temperature:Ambient
CAS Number:112-00-5
Formula:C₁₅H₃₄ClN
MDL Number:MFCD00041974