Order Entry
Germany
ContactUsLinkComponent
Trihexyl(tetradecyl)phosphonium bromide ≥95% (CYPHOS® IL 102)
Trihexyl(tetradecyl)phosphonium bromide ≥95% (CYPHOS® IL 102)
Catalog # STRE15-6378
CAS Number:  
Trihexyl(tetradecyl)phosphonium bromide ≥95% (CYPHOS® IL 102)
Catalog # STRE15-6378
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :563.76 g/mol
Density:0,952
 :GHS07
CAS Number:654057-97-3
Formula:C₃₂H₆₈BrP
MDL Number:MFCD04117697