Dimethyldioctadecylammonium bromide (DDA), Sigma-Aldrich®
Catalog # SIAL40165-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:630.952 g/mol
EINECS:223-037-6
CAS Number:3700-67-2
Formula:C₃₈H₈₀BrN
MDL Number:MFCD00041975